Learn More
Iloperidone metabolite Hydroxy Iloperidone, MedChemExpress
Description
Iloperidone metabolite Hydroxy Iloperidone (P88
Specifications
Specifications
| Chemical Name or Material | Iloperidone metabolite Hydroxy Iloperidone |
| CAS | 133454-55-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H29FN2O4 |
| Quantity | 5 mg |
| Synonym | P88 Hydroxy Iloperidone |
| Solubility Information | DMSO : 25 mg/mL (58.34 mM; Need ultrasonic) |
| SMILES | OC(C)C1=CC=C(OCCCN2CCC(C3=NOC4=C3C=CC(F)=C4)CC2)C(OC)=C1 |
| Molecular Weight (g/mol) | 428.5 |
| Show More |
Research purposes only