missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT1/CD85h/LILRA2 Antibody (600007), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6364PECY7
This item is not returnable.
View return policy
Description
ILT1/CD85h/LILRA2 Monoclonal antibody specifically detects ILT1/CD85h/LILRA2 in Human samples. It is validated for Flow Cytometry
Specifications
| ILT1/CD85h/LILRA2 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11027 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 600007 | |
| Flow Cytometry | |
| CD85 antigen-like family member H, CD85h, CD85h antigen, ILT-1, ILT1CD85H, Leukocyte immunoglobulin-like receptor 7, leukocyte immunoglobulin-like receptor subfamily A member 2, leukocyte immunoglobulin-like receptor subfamily A member 2 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2, LIR7LIR-7Immunoglobulin-like transcript 1 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRA2/CD85h/ILT1, Pro17-Ser437, Accession # Q8N149 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction