missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT2/CD85j/LILRB1 Antibody (VMP55), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50475AF405
This item is not returnable.
View return policy
Description
ILT2/CD85j/LILRB1 Monoclonal specifically detects ILT2/CD85j/LILRB1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| ILT2/CD85j/LILRB1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LILRB1 | |
| Hairy cell leukaemia cells | |
| 0.1 mL | |
| Immunology | |
| 10859 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry | |
| VMP55 | |
| Western Blot, Flow Cytometry | |
| CD85 antigen-like family member J, CD85j, CD85j antigen, Ig-like transcript 2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, Monocyte/macrophage immunoglobulin-like receptor 7 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction