missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT4/CD85d/LILRB2 Antibody (JTX-8064), Alexa Fluor™ 700, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28041AF700
This item is not returnable.
View return policy
Description
ILT4/CD85d/LILRB2 Monoclonal antibody specifically detects ILT4/CD85d/LILRB2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ILT4/CD85d/LILRB2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| CD85 antigen-like family member D, CD85d antigen, Ig-like transcript 4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10 | |
| LILRB2/ ILT4/ CD85d | |
| 0.1 mL | |
| Immunology | |
| 10288 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| JTX-8064 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction