missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT5/CD85a/LILRB3 Antibody (4F9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64329B
This item is not returnable.
View return policy
Description
ILT5/CD85a/LILRB3 Monoclonal specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| ILT5/CD85a/LILRB3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| LILRB3 | |
| Monocyte derived dendritic cells | |
| 0.1 mL | |
| Primary | |
| NB100-64329 is specific for human CD85a (leucocyte immunoglobulin-like receptor, subfamily B, member 3/ LILRB3), also known as ILT-5, expressed by monocytes, dendritic cells, macrophages, subpopulation of T lymphocytes, B cells and at high levels on plasma cells. CD85a is a 631 amino acid type I transmembrane glycoprotein, which contains four immunoreceptor tyrosine-based inhibition motif (ITIM) sequences within a long cytoplasmic tail. Phosphorylation of the tyrosine residues within ITIMs is known to enable the binding and activation of protein tyrosine phosphatases, which act as cell signalling modulators and inhibitors of cell activation. | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 4F9 | |
| Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD85 antigen-like family member A, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 11025 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction