missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT6/CD85e/LILRA3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00150CL1
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
ILT6/CD85e/LILRA3 Polyclonal antibody specifically detects ILT6/CD85e/LILRA3 in Human samples. It is validated for ELISA
Tekniske data
| ILT6/CD85e/LILRA3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 11026 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| CoraFluor 1 | |
| CD85 antigen-like family member E, CD85e antigen, HM31, ILT-6, ILT6CD85e, Immunoglobulin-like transcript 6, leucocyte immunoglobulin-like receptor, Leukocyte immunoglobulin-like receptor 4, leukocyte immunoglobulin-like receptor A3, leukocyte immunoglobulin-like receptor subfamily A member 3, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member3, LIR4CD85E, LIR-4HM43e3, Monocyte inhibitory receptor HM43/HM31 | |
| Produced in rabbits immunized with purified, recombinant Human ILT6/CD85e/LILRA3 (Accession#: AAH28208.1; Met1-Glu439) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion