missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT8/CD85b/LILRA6 Antibody (921330) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8656JF525
This item is not returnable.
View return policy
Description
ILT8/CD85b/LILRA6 Monoclonal antibody specifically detects ILT8/CD85b/LILRA6 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| ILT8/CD85b/LILRA6 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD85b;ILT5;ILT8;ILT-8;LILRB3;LILRB6 | |
| HEK293 human embryonic kidney cell line-derived recombinant human LILRA6/CD85b, Met1-Asn447, Accession # Q6PI73 | |
| RUO | |
| 79168 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 921330 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction