missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILVBL Mouse anti-Human, Rat, Clone: OTI8A12, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503078
This item is not returnable.
View return policy
Description
The protein encoded by this gene shares similarity with several thiamine pyrophosphate-binding proteins identified in bacteria, yeast, and plants. The highest degree of similarity is found with bacterial acetolactate synthases (AHAS), which are enzymes that catalyze the first step in branched-chain amino acid biosynthesis.Specifications
| ILVBL | |
| Monoclonal | |
| 0.56 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| A1L0T0 | |
| ILVBL | |
| Full length human recombit protein of human ILVBL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI8A12 | |
| Unconjugated | |
| ILVBL | |
| 209L8, AHAS, HACL1L, ILV2H | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10994, 362843 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction