Learn More
Imazamox, TRC
Product Code. 17963955
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963955 500 mg 500mg
17931945 50 mg 50mg
17954305 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17963955 Supplier Toronto Research Chemicals Supplier No. TRCI268550500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 114311-32-9
Molecular Formula C15 H19 N3 O4
Molecular Weight (g/mol) 305.33
Synonym Imazamox, 2-[4,5-Dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-(methoxymethyl)-3-pyridinecarboxylic acid, (±)-Imazamox, 2-[4-Isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl]-5-methoxymethylnicotinic acid, 5-Methoxymethyl-2-(4-isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl) nicotinic acid, AC 299263, CL 299263, Global, Imazamox, Pulsar, Pulsar (herbicide), Pulsar 40, Raptor, Raptor (herbicide), Sweeper, Sweeper 70DG, 3-Pyridinecarboxylic acid, 2-[4,5-dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-(methoxymethyl)-
IUPAC Name 5-(methoxymethyl)-2-(4-methyl-5-oxo-4-propan-2-yl-1H-imidazol-2-yl)pyridine-3-carboxylic acid
SMILES COCc1cnc(C2=NC(C)(C(C)C)C(=O)N2)c(c1)C(=O)O
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C15H19N3O4/c1-8(2)15(3)14(21)17-12(18-15)11-10(13(19)20)5-9(6-16-11)7-22-4/h5-6,8H,7H2,1-4H3,(H,19,20)(H,17,18,21)
Formula Weight 305.1376
Percent Purity >95%
Chemical Name or Material Imazamox
Grade HPLC