missing translation for 'onlineSavingsMsg'
Learn More

Iminostilbene-d10, TRC

Product Code. 30405539
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405539 5 mg -
30394174 25 mg 25mg
30398803 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30405539 Supplier Toronto Research Chemicals Supplier No. TRCI4450045MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C14 D10 H N
Molecular Weight (g/mol) 203.305
Synonym 5H-Dibenz[b,f]azepine-d10, 2,2'-Iminostilbene-d10, 5-Azadibenzo[a,e]cycloheptatriene-d10, 5H-Dibenzo[b,f]azepine-d10, NSC 123458-d10, RP 9989-d10
IUPAC Name 1,2,3,4,5,6,7,8,9,10-decadeuterio-11H-benzo[b][1]benzazepine
SMILES [2H]C1=C([2H])c2c([2H])c([2H])c([2H])c([2H])c2Nc3c([2H])c([2H])c([2H])c([2H])c13

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H11N/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)15-13/h1-10,15H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
Formula Weight 203.152
Chemical Name or Material Iminostilbene-d10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.