missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMMP2L Polyclonal Antibody

Description
This antibody is predicted to react with mouse based on sequence homology.
IMP2L complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 and IMP2 are the catalytic subunits of the IMP complex.
Specifications
Specifications
| Antigen | IMMP2L |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | IMMP2L |
| Gene Accession No. | Q96T52 |
| Gene Alias | AI853880; IMMP2L; IMMP2L-IT1; IMP2; IMP2 inner mitochondrial membrane peptidase-like; IMP2 inner mitochondrial membrane peptidase-like (S. cerevisiae); IMP2-LIKE; IMP2-like protein; inner mitochondrial membrane peptidase 2 like; inner mitochondrial membrane peptidase subunit 2; mitochondrial inner membrane protease subunit 2 |
| Gene Symbols | IMMP2L |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?