missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IMP Dehydrogenase 2/IMPDH2 Antibody (SR3949), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27817-100ul
This item is not returnable.
View return policy
Description
IMP Dehydrogenase 2/IMPDH2 Monoclonal antibody specifically detects IMP Dehydrogenase 2/IMPDH2 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| IMP Dehydrogenase 2/IMPDH2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR3949 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| EC 1.1.1.205, IMP (inosine 5'-monophosphate) dehydrogenase 2, IMP (inosine monophosphate) dehydrogenase 2, IMP dehydrogenase 2, IMPD 2, IMPD2IMP oxireductase 2, IMPDH 2, IMPDH-II, inosine 5' phosphate dehydrogenase 2, inosine monophosphate dehydrogenase type II, inosine-5'-monophosphate dehydrogenase 2 | |
| A synthesized peptide derived from human IMP Dehydrogenase 2/IMPDH2 (Uniprot #: P12268) | |
| 100 μL | |
| Lipid and Metabolism | |
| 3615 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction