missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP2/IGF2BP2 Antibody (OTI3F9), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71615H
This item is not returnable.
View return policy
Description
IMP2/IGF2BP2 Monoclonal specifically detects IMP2/IGF2BP2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| IMP2/IGF2BP2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| IGF2BP2 | |
| Full length recombinant protein of human IMP2/IGF2BP2 (NP_006333) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3F9 | |
| Western Blot, Immunohistochemistry | |
| Hepatocellular carcinoma autoantigen p62, IGF-II mRNA-binding protein 2, IMP-2IGF2 mRNA-binding protein 2, IMP2VICKZ family member 2, insulin-like growth factor 2 mRNA binding protein 2, insulin-like growth factor 2 mRNA-binding protein 2, p62, VICKZ2 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 10644.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction