missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP3 Antibody (32E9), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59475AF700
This item is not returnable.
View return policy
Description
IMP3 Monoclonal antibody specifically detects IMP3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| IMP3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| BRMS2mitochondrial ribosomal protein S4, C15orf12, chromosome 15 open reading frame 12, FLJ10968, IMP3, U3 small nucleolar ribonucleoprotein, homolog (yeast), MRPS4DKFZp586L0118, U3 small nucleolar ribonucleoprotein protein IMP3, U3 snoRNP protein 3 homolog, U3 snoRNP protein IMP3 | |
| Recombinant human IMP3 (1-184aa) purified from E. coli | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 55272 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 32000000000 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction