missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111518
This item is not returnable.
View return policy
Description
Immunogen sequence: DFLNWEVTDH NLHELRVLRR YRLQRREDYT RYNQLSRAVR ELARRLRDLP ERDQFRVRAS AALLDKLYAL GLVPT Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
This gene encodes the human homolog of the yeast Imp3 protein. The protein localizes to the nucleoli and interacts with the U3 snoRNP complex. The protein contains an S4 domain.
Specifications
| IMP3 | |
| Polyclonal | |
| Unconjugated | |
| Imp3 | |
| 1190002L16Rik; AI256594; BRMS2; C15orf12; cancer/testis antigen 98; CT98; DKFZp586L0118; FLJ10968; hKOC; IGF2 mRNA-binding protein 3; IGF2BP3; IGF-II mRNA-binding protein 3; IMP3; IMP-3; IMP3, U3 small nucleolar ribonucleoprotein; IMP3, U3 small nucleolar ribonucleoprotein, homolog; IMP3, U3 small nucleolar ribonucleoprotein, homolog (yeast); insulin like growth factor 2 mRNA binding protein 3; insulin-l; insulin-like growth factor 2 mRNA binding protein 3; insulin-like growth factor 2 mRNA-binding protein 3; KH domain containing protein overexpressed in cancer; KH domain-containing protein overexpressed in cancer; KOC; KOC1; MRPS4; OTTHUMP00000183524; RGD1306825; U3 small nucleolar ribonucleoprotein protein IMP3; U3 snoRNP protein 3 homolog; U3 snoRNP protein IMP3; VICKZ family member 3; VICKZ3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55272 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NV31 | |
| Imp3 | |
| Recombinant Protein corresonding to Human IMP3. Recombinant protein control fragment (Product #RP-107045). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction