missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMPACT Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42678B
This item is not returnable.
View return policy
Description
IMPACT Polyclonal antibody specifically detects IMPACT in Human samples. It is validated for Western Blot
Specifications
| IMPACT | |
| Polyclonal | |
| Western Blot | |
| Impact homolog (mouse), Imprinted and ancient gene protein homolog, MGC33718, protein IMPACT | |
| The immunogen for this product maps to a region between residue 270 and 320 of human imprinted and ancient homolog using the numbering given in entry NP_060909.1 (GeneID 55364). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 55364 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction