missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMPDH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116702
This item is not returnable.
View return policy
Description
Positive Samples: BT-474, HT-29, Mouse pancreas Immunogen sequence: MMGSLLAATT EAPGEYFFSD GVRLKKYRGM GSLDAMEKSS SSQKRYFSEG DKVKIAQGVS GSIQDKGSIQ KFVPYLIAGI QHGCQDIGAR SLSVLRSMMY SGELKFEKRT MSAQIEGGVH GLHSYEKRLY.
The protein encoded by this gene acts as a homotetramer to regulate cell growth. The encoded protein is an enzyme that catalyzes the synthesis of xanthine monophosphate (XMP) from inosine-5'-monophosphate (IMP). This is the rate-limiting step in the de novo synthesis of guanine nucleotides. Defects in this gene are a cause of retinitis pigmentosa type 10 (RP10). Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| IMPDH1 | |
| Polyclonal | |
| Unconjugated | |
| IMPDH1 | |
| B930086D20Rik; DKFZp781N0678; HAMAP-Rule:MF_03156}; IMP (inosine 5'-monophosphate) dehydrogenase 1; IMP (inosine monophosphate) dehydrogenase 1; IMP dehydrogenase 1; IMP dehydrogenase 1 {ECO:0000255; IMPD; IMPD 1; IMPD 1 {ECO:0000255; IMPD1; IMPDH 1; IMPDH 1 {ECO:0000255; Impdh1; IMPDH-I; inosine 5'-phosphate dehydrogenase 1; inosine monophosphate dehydrogenase 1; inosine-5'-monophosphate dehydrogenase 1; inosine-5'-monophosphate dehydrogenase 1 {ECO:0000255; LCA11; RP10; sWSS2608 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23917, 3614, 362329 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.05 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D3ZLZ7, P20839, P50096 | |
| IMPDH1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 434-563 of human IMPDH1 (NP_8990661). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction