missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Importin 13 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16627064
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16627064 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16627064 Supplier Invitrogen™ Supplier No. PA5100579

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IPO13.

This gene encodes a member of the importin-beta family of nuclear transport proteins. The encoded protein mediates the import of specific cargo proteins from the cytoplasm to the nucleus and is dependent on the Ras-related nuclear protein-GTPase system. The encoded protein is also involved in nuclear export of the eukaryotic translation initiation factor 1A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Importin 13
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IPO13
Gene Accession No. O94829, Q8K0C1, Q9JM04
Gene Alias Imp13; importin 13; importin-13; Ipo13; Kap13; karyopherin 13; karyopherin-13; KIAA0724; late gestation lung 2; late gestation lung 2 protein; LGL2; Ran binding protein 13; ran-binding protein 13; RanBP13; RP11-7O11.4
Gene Symbols IPO13
Host Species Rabbit
Immunogen A synthesized peptide derived from human IPO13(Accession O94829), corresponding to amino acid residues A636-Q686.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116458, 230673, 9670
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.