missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Importin 7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115423
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Importin 7.
The importin-alpha/beta complex and the GTPase Ran mediate nuclear import of proteins with a classical nuclear localization signal. The protein encoded by this gene is a member of a class of approximately 20 potential Ran targets that share a sequence motif related to the Ran-binding site of importin-beta. Similar to importin-beta, this protein prevents the activation of RaNULLs GTPase by RanGAP1 and inhibits nucleotide exchange on RanGTP, and also binds directly to nuclear pore complexes where it competes for binding sites with importin-beta and transportin. This protein has a Ran-dependent transport cycle and it can cross the nuclear envelope rapidly and in both directions. At least four importin beta-like transport receptors, namely importin beta itself, transportin, RanBP5 and RanBP7, directly bind and import ribosomal proteins.
Specifications
| Importin 7 | |
| Polyclonal | |
| Unconjugated | |
| Ipo7 | |
| A330055O14Rik; C330016G14; FLJ14581; Imp7; importin 7; importin-7; IPO7; MGC138673; RAN binding protein 7; RAN-binding protein 7; RanBP7 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10527, 233726, 308939 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O95373, Q9EPL8 | |
| Ipo7 | |
| A synthesized peptide derived from human IPO7(Accession O95373), corresponding to amino acid residues R1003-N1038. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction