missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin-9 Antibody (SR1154), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22553-100ul
This item is not returnable.
View return policy
Description
Importin-9 Monoclonal antibody specifically detects Importin-9 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Importin-9 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1154 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| DKFZp761M1547, FLJ10402, Imp9IMP9, importin 9, importin-9, KIAA1192, Ran-binding protein 9, RANBP9 | |
| A synthesized peptide derived from human Importin-9 (Uniprot #: Q96P70) | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 55705 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction