missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin beta/KPNB1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79806AF532
This item is not returnable.
View return policy
Description
Importin beta/KPNB1 Polyclonal antibody specifically detects Importin beta/KPNB1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Importin beta/KPNB1 | |
| Polyclonal | |
| Western Blot | |
| IMB1, Impnb, importin 1, importin 90, importin beta-1 subunit, importin subunit beta-1, importin-90, IPO1, IPOB, karyopherin (importin) beta 1, Karyopherin subunit beta-1, MGC2155, MGC2156, MGC2157, NTF97Pore targeting complex 97 kDa subunit, Nuclear factor p97, PTAC97 | |
| The immunogen recognized by this antibody maps to a region between residue 825 and the C-terminus (residue 876) of human Karyopherin (importin) beta 1 using the numbering given in entry NP_002256.2 (GeneID 3837). | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 3837 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction