missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INAVA Antibody (OTI6A6), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72021R
This item is not returnable.
View return policy
Description
INAVA Monoclonal specifically detects INAVA in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| INAVA | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| C1ORF106 | |
| Recombinant protein fragment corresponding to amino acids 1-270 of human C1orf106 (NP_060735) produced in E.coli. | |
| 0.1 mL | |
| 55765.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI6A6 | |
| Western Blot, Immunohistochemistry | |
| C1orf106, chromosome 1 open reading frame 106, hypothetical protein LOC55765 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction