missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ INCA-6

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
INCA-6 is an inhibitor of interaction between calcineurin and its substrate nuclear factor of activated T cells (NFAT); blocks at the substrate recognition site but not at the catalytic site (Kd = 0.8 mM). Inhibits NFAT dephosphorylation and nuclear import. Also prevents induction of cytokine mRNAs that are downstream targets of NFAT.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | INCA-6 |
| Purity | >99% |
| Molecular Weight (g/mol) | 284.31 |
| Inhibitors | INCA-6 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction