missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX InCELL Hunter™ eXpress HEK 293 BRDT(1,2) Bromodomain Assay
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Description
The InCELL Hunter™ eXpress HEK 293 BRDT(1,2) Bromodomain Assay provides an easy-to-use cell based assay to study small molecule drug candidates that engage with regulators of epigenetics pathways. This kit assesses compound-engagement-based stabilization of BRDT(1,2) via detection of BRDT(1,2) levels. This product uses EFC technology. The cell line included overexpresses BRDT(1,2) target protein fused to PL. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | HEK 293 |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | (+)-JQ1 |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | BRDT(1,2) |
| Cell Line | 293H |
| Target | BRDT(1,2) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction