missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Inclacumab Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA541955
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Inclacumab has been used in trials studying the treatment and prevention of Myocardial Infarction, Peripheral Arterial Disease (PAD), and Coronary Heart Disease, Graft Occlusion, Vascular.
Specifications
| Inclacumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| LC1004-002; RO4905417 | |
| Human SELP. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 1.68 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG4 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction