Learn More
PEPperCHIP™ Incubation Tray 3/5
Click to view available options
Number of Arrays:
3 arrays
Unit Size:
Each
Description
- Minimal sample consumption
- Shaking incubation enabled
- Fully reusable
- Components: PVC-C base plate, PVC-C tray well plate with screws, silicon seal and acryl lid
- Tray size: 12.8 x 8.55 cm
- Well size: 5, 11.6 x 20 mm
Specifications
Specifications
| Number of Arrays | 3 arrays |
| Format | 3 slots, 3 PEPperCHIP Peptide Microarrays, Format 5 |
| Color | Black |
| Content And Storage | Room Temperature |
| For Use With (Application) | Antibody validation, epitope mapping, serum screening, biomarker discovery, protein-peptide interaction studies |
| For Use With (Equipment) | PEPperCHIP™ Peptide Microarrays |
| Includes | Base plate, slide format specific well plate and seal, screws and lid cover |
| No. of Samples | 15 |