Learn More
MMM Medcenter™ FRIOCELL™ Refrigerated Incubator, 55 L
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
55 L
missing translation for 'unitSize'
Each
Specifications
Specifications
| Capacity (Metric) Shelf | 20 kg |
| Capacity (Metric) | 55 L |
| Lighting | Phytoculture |
| No. of Shelves | 4 |
| Temperature Range (Metric) | 0°C to 99.9°C |
| Display | Digital |
| Temperature Stability | ± 0.2°C |
| Height (Metric) Interior | 35 cm |
| Depth (Metric) Interior | 37 cm |
| Weight (Metric) | 80 kg |
| Show More |