Learn More
Aesculap™ Indicator Label
Product Code. 15757722
Click to view available options
Dimensions (L x W):
67 x 40 mm
Unit Size:
Pack of 1000
Product Code. 15757722
Brand: Aesculap™ JG682
Only null left
Add to basket
Add to basket
Aesculap™ Indicator Label with process indicator for steam sterilization of legacy standard sterile containers.
  • Indicator label with process indicator for steam sterilization
  • For legacy standard sterile containers
Specifications
Dimensions (L x W) 67 x 40 mm
Product Type Indicator Label
Quantity 1000/Pk.