Learn More
Macherey-Nagel™ Special pH Indicator Paper Reel
Product Code. 10414851 Shop All Macherey-Nagel Products
Click to view available options
pH Range:
8 to 10
Unit Size:
5m
Product Code. 10414851
Brand: Macherey-Nagel™ 90212
Only null left
Add to basket
Add to basket
The indicator paper comes in a plastic reel that ensures long-term stability and protection against many external influences.
  • Gradation: 8.0, 8.2, 8.4, 8.7, 9.0, 9.2, 9.6, 10.0
  • Evaluable on reflectometer: No
Specifications
pH Range 8 to 10
Quantity 1 Each
Storage Requirements 4°C to 30°C
Length (Metric) 5 m
Width (Metric) 7 mm