missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Indiko™ System Calibrators and Controls

Product Code. 10262285 Shop All Thermo Scientific Products
Change view
Click to view available options
Detectable Analytes:
uTrol Control
uTrol Control, High
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes unitSize
10262285 uTrol Control Pack of 5
10528505 uTrol Control, High Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10262285 Supplier Thermo Scientific™ Supplier No. 981821
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Enable accurate test results with Thermo Scientific™ Indiko™ System Calibrators and Controls.

Enable accurate results with Thermo Scientific™ Indiko™ System Calibrators and Controls. Indiko analyzers are fully automated for routine and specialty testing. Ready-to-use barcoded reagents are designed to support efficient operations and reduce the risk of errors.

Highlights:
  • Based on serum or urine of human or bovine origin
  • Provided in a lyophilized form
Ordering information:
  • Nortrol, Abtrol and sCal are available in the U.S.A.
  • All other calibrators and controls are not available in the U.S.A.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Compliant with the IVDR; CE marked
Detectable Analytes uTrol Control
Sample Type Urine
Type Control
Quantity 10 x 5 mL
compliance-icons
Product Identifier
  • X5 uTrol control, 5 x 10ml
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.