Learn More
Thermo Scientific™ Indiko™ System Calibrators and Controls
Product Code. 10528505 Shop All Thermo Scientific Products
Change view
Click to view available options
Detectable Analytes:
uTrol Control
uTrol Control, High
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
10528505 uTrol Control, High Pack of 5
10262285 uTrol Control Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10528505 Supplier Thermo Scientific™ Supplier No. 981822
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Enable accurate test results with Thermo Scientific™ Indiko™ System Calibrators and Controls.
Enable accurate results with Thermo Scientific™ Indiko™ System Calibrators and Controls. Indiko analyzers are fully automated for routine and specialty testing. Ready-to-use barcoded reagents are designed to support efficient operations and reduce the risk of errors.
Highlights:
  • Based on serum or urine of human or bovine origin
  • Provided in a lyophilized form
Ordering information:
  • Nortrol, Abtrol and sCal are available in the U.S.A.
  • All other calibrators and controls are not available in the U.S.A.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Compliant with the IVDR; CE marked
Detectable Analytes uTrol Control, High
Sample Type Urine
Type Control
Quantity 10 x 5 mL