missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents, GGT to Uric Acid

Product Code. 10606557 Shop All Thermo Scientific Products
Change view
Click to view available options
Detectable Analytes:
U/CSF Protein
Quantity:
8 x 20 mL
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Quantity Unit Size
10606557 U/CSF Protein 8 x 20 mL Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10606557 Supplier Thermo Scientific™ Supplier No. 981843
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fully standardize performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents.

Enhance your laboratory's performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents. These ready-to-use assays come fully validated and compliant, featuring automatic barcoding for efficient process and reordering checks. System reagents provide timely and accurate information, helping to ensure results you can stand behind.

  • Indiko™ and Konelab™ applications utilize carefully selected methods according to IFCC and other widely used recommendations
  • Clinical chemistry methods are provided with system specified multi-calibrators and -controls with traceability to international reference materials
  • CE marked kits are compliant with the IVDD
Ordering Information: Not available in the U.S.A.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Description U/CSF Protein; Kit Size: 8 x 20mL
CE Marker Yes
Detectable Analytes U/CSF Protein
Sample Type Serum
Type Reagent
Quantity 8 x 20 mL
Form Liquid
compliance-icons
Product Identifier
  • U/CSF Protein reagent 8x 20ml
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid, .alpha.-[4-(1,1,3,3-tétraméthylbutyl)phényl]-.oméga.-hydro xy-Poly(oxy-1,2-éthanediy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.