Learn More
Thermo Scientific™ Indiko™ System D-Dimer to Transferrin Specific Protein Reagents
Product Code. 10038254 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Reagent
Quantity:
2 x 3 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity Unit Size
10038254 Reagent 2 x 3 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10038254 Supplier Thermo Scientific™ Supplier No. 981935
  • ONLINE EXCLUSIVE PRICE
    € 174.00
    € 156.60 / Pack of 1
    Save € 17.40
    10% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Choose from a specific protein test menu of over 20 immunoturbidimetric methods with Thermo Scientific™ Indiko™ System Specific Protein Reagents.
Receive best-in-class methodologies and standardized performance with Thermo Scientific™ Indiko™ System D-Dimer to Transferrin Specific Protein Reagents, which include a specific protein test menu of over 20 immunoturbidimetric methods. The assays are always ready to go thanks to full validation and compliance as well as barcoding.
All Konelab and Indiko methods are based on carefully-designed protocols for barcoded system reagents. All methods are provided, and calibrators and controls are available in separate kits. CE marked kits are compliant with the IVDD.
Ordering information: Not available in the U.S.A.
Specific Protein Applications for Indiko™ and Konelab™ Analyzers:
  • Albumin / Microalbuminuria
  • Prealbumin
  • Antistreptolysin O
  • Alpha-1-Antitrypsin
  • Apolipoprotein A1 & B
  • Ceruloplasmin
  • Complement C3 & C4
  • C-reactive protein
  • Cystatin C
  • D-Dimer
  • Ferritin
  • HbA1c
  • Haptoglobin
  • IgA, G & M
  • Lipoprotein(a)
  • Myoglobin
  • Orosomucoid
  • Rheumatoid factors
  • Soluble Transferrin Receptor
  • Transferrin
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Marked
CE Marker Yes
Detectable Analytes Haptoglobin
Sample Type Serum
Type Reagent
Quantity 2 x 3 mL
Form Liquid