Learn More
Indoleamine 2,3-dioxygenase/IDO Antibody (700838), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB6030Z
Product Code. 18528886
  • € 712.00 / 0.10mL
Description
Indoleamine 2,3-dioxygenase/IDO Monoclonal antibody specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Indoleamine 2,3-dioxygenase/IDO
Monoclonal
DyLight 755
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
700838
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase
E. coli-derived recombinant human Indoleamine 2,3-dioxygenase/IDO, Ala2-Gly403, Accession # P14902
0.1 mL
Cancer, Myeloid derived Suppressor Cell
3620
Store at 4°C in the dark.