Learn More
3M™ Maintenance Sorbent Pads M-A2002
Product Code. 10757835
Click to view available options
:
Industrial Absorbent Sheet
Product Code. 10757835
Brand: 3M™ 7100003876
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Maintenance Sorbent Pads are designed to be used in and around the work area to catch leaks and drips, pads are highly absorbent and simple to use.
  • Sorbent pads designed for use in and around the work area to catch leaks and drips
  • For use on most non-aggressive liquids
  • Very easy to use and to retrieve
  • Promotes a cleaner, safer working environment
Order Info
dimProductType
TRUSTED_SUSTAINABILITY
Specifications
Color Grey
Type Sorbent Pads
Length (Metric) 520 mm
Width (Metric) 400 mm