Learn More
Tocris Bioscience™ INF 4E
Click to view available options
:
10mg
Description
INF 4E is a NLRP3 inflammasome and caspase-1 inhibitor (Ki = 9.6 μM for caspase-1). Inhibits Nigericin (Cat. No. 4312) or ATP (Cat.No. 3245) induced pyroptosis of PMA-differentiated and LPS-primed THP-1 cells and activates the Keap1-Nrf2 pathway.
Specifications
Specifications
| Chemical Name or Material | Ethyl 2[(2-chlorophenyl)(hydroxy)methyl]acrylate |
| CAS | 88039-46-7 |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Molecular Formula | C12H13ClO3 |
| Formula Weight | Observed MW: 240.68 |