missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79025
This item is not returnable.
View return policy
Description
INF2 Polyclonal specifically detects INF2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| INF2 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| C14orf151, C14orf173DKFZp762A0214, chromosome 14 open reading frame 151, chromosome 14 open reading frame 173, FLJ22056, FSGS5, HBEAG-binding protein 2 binding protein C, HBEBP2-binding protein C, inverted formin, FH2 and WH2 domain containing, inverted formin-2, MGC13251, pp9484 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 64423 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate | |
| NP_071934.3 | |
| INF2 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Inverted Formin-2 using the numbering given in entry NP_071934.3 (GeneID 64423). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction