missing translation for 'onlineSavingsMsg'
Learn More

INF2 Mouse anti-Human, Unconjugated, Clone: 4A11A7, Proteintech

Product Code. p-7223586 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16889195 20 μL 20µL
16879195 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16889195 Supplier Proteintech Supplier No. 669101IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The INF2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human INF2. This antibody recognizes human antigen. The INF2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antigen INF2
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 4A11A7
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 64423
Target Species Human
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.