Learn More
Infigratinib, MedChemExpress
Product Code. 16448855
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448855 10 mg 10mg
16441076 200 mg 200mg
16431807 50 mg 50mg
16416166 100 mg 100mg
16408337 5 mg 5mg
16401666 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16448855 Supplier Medchem Express Supplier No. HY13311
MedChemExpress Infigratinib (BGJ-398; NVP-BGJ398) is a potent inhibitor of the FGFR family with IC50s of 0.9 nM, 1.4 nM, 1 nM, and 60 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively.
Infigratinib (BGJ-398
Specifications
Chemical Name or Material Infigratinib
CAS 872511-34-7
Color White
Purity Grade Notes Research
Molecular Formula C26H31Cl2N7O3
Quantity 10 mg
Synonym BGJ-398 NVP-BGJ398
Solubility Information DMSO : 12 mg/mL (21.41 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES ClC1=C(C(Cl)=C(C=C1OC)OC)NC(N(C2=CC(NC3=CC=C(C=C3)N4CCN(CC4)CC)=NC=N2)C)=O
Molecular Weight (g/mol) 560.48
Formula Weight 560.48
Percent Purity 98.69%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only