missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Influenza A H17N10 HA (A/little yellow-shouldered bat/Guatemala/164/2009) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16388734
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16388734 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16388734 Supplier Invitrogen™ Supplier No. PA581753

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza A H17N10 (A/little yellow-shouldered bat/Guatemala/164/2009) Hemagglutinin.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H17N10 HA (A/little yellow-shouldered bat/Guatemala/164/2009)
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias flu hemagglutinin; influenza hemagglutinin
Host Species Rabbit
Immunogen Recombinant Influenza A H17N10 (A/little yellow-shouldered bat/Guatemala/164/2009) Hemagglutinin Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.