missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 Hemagglutinin Antibody (10), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06578CL1
This item is not returnable.
View return policy
Description
Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H1N1 Hemagglutinin in Influenza A virus H1N1 samples. It is validated for Western Blot
Specifications
| Influenza A H1N1 Hemagglutinin | |
| Monoclonal | |
| CoraFluor 1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Influenza A virus H1N1 | |
| Purified |
| Western Blot | |
| 10 | |
| PBS | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H1N1 Hemagglutinin (A/California/04/2009) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction