missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 Hemagglutinin Antibody (IV.C102), Alexa Fluor™ 488 - (A/USSR/90/77), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65047AF488
This item is not returnable.
View return policy
Description
Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H1N1 Hemagglutinin in Influenza A Virus H1N1 samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).
Specifications
| Influenza A H1N1 Hemagglutinin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HA | |
| Influenza A/USSR/90/77 | |
| 0.1 mL | |
| Primary | |
| Influenza A virus H1N1, Virus | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| IV.C102 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Sandwich ELISA | |
| HA, HA1, HA2, Hemagglutinin, Influenza A Virus Hemagglutinin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| NB100-65047 is specific for the haemagglutinin of the Influenza A H1N1 serotype and does not react with H2N2, H3N2, Influenza B, RSV, Parainfluenza I, II or III, Adenovirus, mumps or measle antigens. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction