missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 M1 Antibody (02), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06633AF350
This item is not returnable.
View return policy
Description
Influenza A H1N1 M1 Monoclonal antibody specifically detects Influenza A H1N1 M1 in Influenza virus N1 samples. It is validated for ELISA
Specifications
| Influenza A H1N1 M1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant H1N1 (A/Puerto Rico/8/34/Mount Sinai) M1/ Matrix Protein 1 (NP_040978.1; Ser 2-Lys 252). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 02 | |
| ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Influenza virus N1 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction