missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 Neuraminidase Antibody (2F10E12G1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06579B
This item is not returnable.
View return policy
Description
Influenza A H1N1 Neuraminidase Monoclonal antibody specifically detects Influenza A H1N1 Neuraminidase in Influenza virus N1 samples. It is validated for Western Blot, ELISA
Specifications
| Influenza A H1N1 Neuraminidase | |
| Monoclonal | |
| Biotin | |
| PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant influenza A virus H1N1 Neuraminidase. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 2F10E12G1 | |
| Western Blot, ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Influenza virus N1 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction