missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 NS1 Antibody - (A/WSN/1933), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-42871
This item is not returnable.
View return policy
Description
Specifically detects Influenza A Virus NS1 in Human, Virus samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting
Influenza A H1N1 NS1 Polyclonal antibody specifically detects Influenza A H1N1 NS1 in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| Influenza A H1N1 NS1 | |
| Polyclonal | |
| Western Blot 1:500-1:3000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin | |
| Influenza NS1 | |
| Recombinant protein of Influenza A virus NS1 (nonstructural protein) (A/WSN/1933(H1N1). The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction