missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 Nucleoprotein Antibody (2F205), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24871AF488
This item is not returnable.
View return policy
Description
Influenza A nucleoprotein Monoclonal specifically detects Influenza A nucleoprotein in Virus samples. It is validated for ELISA.
Specifications
| Influenza A nucleoprotein | |
| Monoclonal | |
| Alexa Fluor 488 | |
| O73619 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 394335 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA | |
| 2F205 | |
| ELISA | |
| Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein | |
| Recombinant influenza A protein was used as the immunogen | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction