missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 PA Antibody - (A/WSN/1933), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-42874
This item is not returnable.
View return policy
Description
Specifically detects Influenza A Virus PA in Virus samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting
Influenza A H1N1 PA Polyclonal antibody specifically detects Influenza A H1N1 PA in Virus samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
| Influenza A H1N1 PA | |
| Polyclonal | |
| Western Blot 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Influenza PA | |
| Recombinant protein encompassing a sequence within the C-terminus region of Influenza A virus PA (A/WSN/1933(H1N1)). The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Human, Virus | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| Rabbit | |
| 83 kDa | |
| 0.1 mL | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction