missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H5N1 Hemagglutinin Mouse anti-Virus, Alexa Fluor 532, Clone: 1E7D8, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-75494AF532
This item is not returnable.
View return policy
Description
Influenza A H5N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Influenza A H5N1 Hemagglutinin | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, ELISA | |
| 1E7D8 | |
| 50mM Sodium Borate | |
| A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein. | |
| 0.1 mL | |
| Immunology, Virology Bacteria and Parasites | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction