Learn More
Influenza A H6N1 Hemagglutinin Antibody (8A10D8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06474-100ul
This item is not returnable.
View return policy
Description
Influenza A H6N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H6N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Influenza A H6N1 Hemagglutinin | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A virus H6N1 Hemagglutinin. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 8A10D8 | |
| Western Blot 1:500-1:1000, ELISA 1:1000-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |