missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A H6N1 MP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141124
This item is not returnable.
View return policy
Description
This sequence is 100% conserved in chicken, duck, and porcine Influenza A.
An H6N1 influenza virus was isolated from a green-winged teal during the H5N1 outbreak in Hong Kong Special Administrative Region (SAR) in 1997. This virus possesses similar genes encoding internal proteins as in the human H5N1 and H9N2 influenza viruses. In 1999, influenza viruses from quail infected two humans in Hong Kong. suggesting the potential for avian influenza viruses to cross the species barrier and infect humans without prior reassortment in an intermediate host, such as the pig. The common features shared by H5N1 and H9N2 influenza viruses isolated from humans are the genes encoding the proteins of the replicating complex, the matrix protein (M) gene, the nonstructural protein (NS) gene, N1 neuraminidase (NA), This virus essentially represents the reemergence of the H5N1 influenza viruses with a different hemagglutinin (HA).
Specifications
| Influenza A H6N1 MP1 | |
| Polyclonal | |
| Unconjugated | |
| H6N1 MP1; Influenza H6N1 | |
| Amino acids 9-28 of avian flu matrix protein 1. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Rabbit | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction